Tetrabromo-p-benzoquinone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,3,5,6-Tetrabromo-1,4-benzoquinone, 2,3,5,6-Tetrabromocyclohexa-2,5-diene-1,4-dione, p-Bromanil, Tetrabromoquinone, 4-Bromanil, Tetrabromo-1,4-benzoquinone
A481699-1G
A481699-25G
A481699-5G
A481699-100G
76695-536EA
10
USD
76695-536
76695-538
76695-540
77626-958
Tetrabromo-p-benzoquinone 98%
Tetrabromo-p-benzoquinone
Precaución: for Research Use Only
Formula:
C₆Br₄O₂ MW: 423.68 g/mol |
MDL Number:
MFCD00013785 CAS Number: 488-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...