Cesium bromide 99%

Supplier: Ambeed
Warning

A481379-100G A481379-10G A481379-25G A481379-500G A481379-5G
76695-524EA 47 USD
76695-524 76695-526 76695-528 76695-530 76695-532
Cesium bromide 99%
Cesium bromide
Formula: BrCs
MDL Number: MFCD00010954
CAS Number: 7787-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR