(R)-3,4-Diamino-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Asp-NH₂

A481238-1G A481238-250MG
76695-520EA 29.71 USD
76695-520 76695-522
(R)-3,4-Diamino-4-oxobutanoic acid 98%
(R)-3,4-Diamino-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₄H₈N₂O₃
MW: 132.119 g/mol
MDL Number: MFCD00237411
CAS Number: 200260-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR