8-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 97%

Supplier: Ambeed
Warning

A481214-10G A481214-5G
76695-516EA 192.03 USD
76695-516 76695-518
8-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 97%
8-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃NO
MW: 292.05 g/mol
MDL Number: MFCD00174291
CAS Number: 59108-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR