7-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 95%

Supplier: Ambeed
Warning

A479876-100MG A479876-1G A479876-250MG A479876-5G
76695-508EA 11.67 USD
76695-508 76695-510 76695-512 76695-514
7-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 95%
7-(Trifluoromethyl)imidazo[1,2-a]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₄F₃N₃
MW: 187.122 g/mol
MDL Number: MFCD08234997
CAS Number: 375857-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR