Cesium iodide 95%

Supplier: Ambeed
Warning

A479132-100G A479132-25G A479132-500G A479132-5G
76695-500EA 83 USD
76695-500 76695-502 76695-504 76695-506
Cesium iodide 95%
Cesium iodide

Precaución: for Research Use Only

Formula: CsI
MDL Number: MFCD00010962
CAS Number: 7789-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR