4-Iodo-1,2-dimethoxybenzene 97%

Supplier: Ambeed
Warning

A478247-100G A478247-10G A478247-1G A478247-25G A478247-5G
76695-480EA 243 USD
76695-480 76695-482 76695-484 76695-486 76695-488
4-Iodo-1,2-dimethoxybenzene 97%
4-Iodo-1,2-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉IO₂
MDL Number: MFCD00094416
CAS Number: 5460-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR