Citral 98%

Supplier: Ambeed
Warning

Synonyms: 3,7-Dimethylocta-2,6-dienal

A477712-25G A477712-500G A477712-1000G
76695-474EA 12 USD
76695-474 76695-476 77551-452
Citral 98%
Citral

Precaución: for Research Use Only

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 229
Flash Pt: 98
MDL Number: MFCD00006997
CAS Number: 5392-40-5
Merck Index: 13,02346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR