1,1-Cyclobutanedimethanol 95%

Supplier: Ambeed
Danger

A477041-10G A477041-1G A477041-5G A477041-250MG
76695-466EA 130 USD
76695-466 76695-468 76695-470 77880-038
1,1-Cyclobutanedimethanol 95%
1,1-Cyclobutanedimethanol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MDL Number: MFCD00463706
CAS Number: 4415-73-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR