N,N-Bis(isopropyl)benzylamine 98%

Supplier: Ambeed
Warning

Synonyms: N-Benzyl-N-isopropylpropan-2-amine, N-Benzyl-N-(prop-2-yl)propan-2-amine

A476647-1G A476647-5G
76695-462EA 20.16 USD
76695-462 76695-464
N,N-Bis(isopropyl)benzylamine 98%
N,N-Bis(isopropyl)benzylamine

Precaución: for Research Use Only

Formula: C₁₃H₂₁N
MW: 191.313 g/mol
MDL Number: MFCD07368226
CAS Number: 34636-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR