Barium trifluoromethanesulphonate 98%

Supplier: Ambeed
Warning

Synonyms: Barium Bis(trifluoromethanesulphonate), Barium triflate, Barium(II) trifluoromethanesulphonate, Trifluoromethanesulphonic acid Barium(II) salt, Ba(OTf)2, Barium(II) triflate

A472227-25G A472227-5G
76695-442EA 34 USD
76695-442 76695-444
Barium trifluoromethanesulphonate 98%
Barium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₂BaF₆O₆S₂
MW: 435.45 g/mol
MDL Number: MFCD00143644
CAS Number: 2794-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR