Allura red AC 85%

Supplier: Ambeed
Warning

Synonyms: Disodium 6-hydroxy-5-[(2-methoxy-5-methyl-4-sulfophenyl)azo]-2-naphthalenesulfonate, FD and C Red No.40, Sodium 6-hydroxy-5-((2-methoxy-5-methyl-4-sulphonatophenyl)diazenyl)naphthalene-2-sulphonate, C.I. Food Red 17, Red No. 40, Allura Red, FD and C Red No.40 (FD&C Red 40), C. I. 16035, Disodium 6-hydroxy-5-[(2-methoxy-5-methyl-4-sulphophenyl)azo]-2-naphthalenesulphonate, Allura Red AC

A471268-1000G A471268-100G A471268-25G A471268-500G
76695-434EA 185 USD
76695-434 76695-436 76695-438 76695-440
Allura red AC 85%
Allura red AC

Precaución: for Research Use Only

Formula: C₁₈H₁₄N₂O₈S₂·2Na
MDL Number: MFCD00059526
CAS Number: 25956-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR