Cinepazide Maleate 98%

Supplier: Ambeed
Warning

Synonyms: 1-[(1-Pyrrolidinylcarbonyl)methyl]-4-(3,4,5-trimethoxycinnamoyl)piperazine Maleate, Vasodistal, 1-(4-(2-Oxo-2-(pyrrolidin-1-yl)ethyl)piperazin-1-yl)-3-(3,4,5-trimethoxyphenyl)prop-2-en-1-one maleate

A469349-10MG A469349-50MG
76695-428EA 5 USD
76695-428 76695-430 76695-432
Cinepazide Maleate 98%
Cinepazide Maleate

Precaución: for Research Use Only

Formula: C₂₂H₃₁N₃O₅·C₄H₄O₄
MW: 533.58 g/mol
MDL Number: MFCD01320526
CAS Number: 26328-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR