Bis(tri-tert-butylphosphine)platinum(0) 98%

Supplier: Ambeed
Warning

Synonyms: Bis(tri-t-butylphosphine)platinum (0)

A466326-100MG A466326-250MG A466326-50MG
76695-402EA 123.07 USD
76695-402 76695-404 76695-406
Bis(tri-tert-butylphosphine)platinum(0) 98%
Bis(tri-tert-butylphosphine)platinum(0)

Precaución: for Research Use Only

Formula: C₂₄H₅₄P₂Pt
MW: 599.73 g/mol
MDL Number: MFCD04974248
CAS Number: 60648-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR