Bis(hexafluoroacetylacetonato)cobalt(II) 98%

Supplier: Ambeed
Warning

A465719-1G A465719-250MG A465719-5G
76695-396EA 25 USD
76695-396 76695-398 76695-400
Bis(hexafluoroacetylacetonato)cobalt(II) 98%
Bis(hexafluoroacetylacetonato)cobalt(II)

Precaución: for Research Use Only

MDL Number: MFCD00042510
CAS Number: 19648-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR