Anthraquinone-2-sulfonic acid sodium salt 97%
Supplier: Ambeed
|
Warning
|
A464915-25G
A464915-5G
76695-384EA
14
USD
76695-384
76695-386
Anthraquinone-2-sulfonic acid sodium salt 97%
Anthraquinone-2-sulfonic acid sodium salt
Precaución: for Research Use Only
Formula:
C₁₄H₇NaO₅S |
MDL Number:
MFCD00001229 CAS Number: 131-08-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...