9H-Fluorene-9,9-dimethanol 98%

Supplier: Ambeed
Warning

A462875-1G A462875-250MG A462875-5G
76695-366EA 15.92 USD
76695-366 76695-368 76695-370 76695-372
9H-Fluorene-9,9-dimethanol 98%
9H-Fluorene-9,9-dimethanol

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MDL Number: MFCD01321348
CAS Number: 4425-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR