(S)-(+)-4-Phenyl-2-oxazolidinone 95%

Supplier: Ambeed
Warning

A462404-1000G A462404-100G A462404-10G A462404-25G A462404-500G A462404-5G
76695-354EA 247 USD
76695-354 76695-356 76695-358 76695-360 76695-362 76695-364
(S)-(+)-4-Phenyl-2-oxazolidinone 95%
(S)-(+)-4-Phenyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD00043396
CAS Number: 99395-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR