Pigment Orange 43 94%

Supplier: Ambeed
Danger

Synonyms: Benzo[lmn]benzo[4,5]imidazo[2,1-b]benzo[4,5]imidazo[2,1-i][3,8]phenanthroline-8,17-dione, Vat Orange 7

A462214-1G A462214-25G A462214-5G
76695-342EA 21 USD
76695-342 76695-344 76695-346
Pigment Orange 43 94%
Pigment Orange 43

Precaución: for Research Use Only

Formula: C₂₆H₁₂N₄O₂
MDL Number: MFCD00071829
CAS Number: 4424-06-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR