(S)-tert-Butyl 2-hydroxy-3-phenylpropanoate 96%

Supplier: Ambeed
Warning

A462081-1G A462081-250MG
76695-338EA 136.86 USD
76695-338 76695-340
(S)-tert-Butyl 2-hydroxy-3-phenylpropanoate 96%
(S)-tert-Butyl 2-hydroxy-3-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
MDL Number: MFCD11042373
CAS Number: 7622-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR