4-Methoxy-2,5-dimethylbenzaldehyde 98%

Supplier: Ambeed
Warning

A459854-1G A459854-5G
76695-320EA 16.98 USD
76695-320 76695-322
4-Methoxy-2,5-dimethylbenzaldehyde 98%
4-Methoxy-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
MDL Number: MFCD00003343
CAS Number: 6745-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR