3-Fluoro-DL-phenylglycine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-(3-fluorophenyl)acetic acid

A459350-100MG A459350-1G A459350-250MG A459350-5G
76695-312EA 6 USD
76695-312 76695-314 76695-316 76695-318
3-Fluoro-DL-phenylglycine 98%
3-Fluoro-DL-phenylglycine

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.16 g/mol
MDL Number: MFCD00049325
CAS Number: 7292-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR