Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) 98%

Supplier: Ambeed
Warning

Synonyms: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl

A459084-10MG A459084-1MG A459084-100MG A459084-250MG A459084-50MG
76695-306EA 22.28 USD
76695-306 76695-308 77626-910 77626-912 77626-914
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) 98%
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

Precaución: for Research Use Only

Formula: C₁₆H₂₅ClN₂O₄
MW: 308.4 · 36.5 g/mol
MDL Number: MFCD00058004
CAS Number: 65391-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR