Acrylic anhydride 98% stabilised

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A457066-1G A457066-25G A457066-5G
76695-284EA 5 USD
76695-284 76695-286 77551-390
Acrylic anhydride 98% stabilised
Acrylic anhydride

Precaución: for Research Use Only

Formula: C₆H₆O₃
MDL Number: MFCD00048146
CAS Number: 2051-76-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR