β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 98%
Supplier: Ambeed
|
Warning
|
A454463-100MG
A454463-1G
A454463-5G
76695-272EA
14.86
USD
76695-272
76695-274
76695-276
76695-278
76695-280
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 98%
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Precaución: for Research Use Only
Formula:
C₂₁H₂₇N₇Na₂O₁₄P₂ |
MDL Number:
MFCD00036200 CAS Number: 606-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...