Oxepane-2,7-dione 90%

Supplier: Ambeed
Warning

A453303-1G A453303-25G A453303-5G A453303-100G A453303-250MG
76695-252EA 18 USD
76695-252 76695-254 76695-256 77878-212 77878-213
Oxepane-2,7-dione 90%
Oxepane-2,7-dione

Precaución: for Research Use Only

Formula: C₆H₈O₃
MDL Number: MFCD01593588
CAS Number: 2035-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR