Bis (8-hydroxyquinolinium) sulfate monohydrate 98%

Supplier: Ambeed
Warning

A452674-100G A452674-25G A452674-500G
76695-246EA 16 USD
76695-246 76695-248 76695-250
Bis (8-hydroxyquinolinium) sulfate monohydrate 98%
Bis(8-hydroxyquinolinium)sulphate monohydrate

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O₆S·1H₂O
MDL Number: MFCD00149610
CAS Number: 207386-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR