2-Amino-2-(3,4-dimethoxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A451587-1G A451587-250MG
76695-242EA 256 USD
76695-242 76695-244
2-Amino-2-(3,4-dimethoxyphenyl)acetic acid 95%
2-Amino-2-(3,4-dimethoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MDL Number: MFCD02662417
CAS Number: 91819-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR