4-Biphenyl benzoate 99%

Supplier: Ambeed
Warning

Synonyms: Benzoic Acid 4-Biphenyl Ester, (4-Phenylphenyl) benzoate

A451349-1G A451349-5G
76695-238EA 5 USD
76695-238 76695-240
4-Biphenyl benzoate 99%
4-Biphenyl benzoate

Precaución: for Research Use Only

Formula: C₁₉H₁₄O₂
MW: 274.32 g/mol
MDL Number: MFCD00223541
CAS Number: 2170-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR