Homophthalic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Carboxymethyl)benzoic acid
A449687-100G
A449687-10G
A449687-25G
A449687-500G
A449687-5G
76695-228EA
81
USD
76695-228
76695-230
76695-232
76695-234
76695-236
Homophthalic acid 97%
Homophthalic acid
Precaución: for Research Use Only
Formula:
C₉H₈O₄ |
MDL Number:
MFCD00004326 CAS Number: 89-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...