(2-Ethoxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A448871-25G A448871-5G A448871-1G
76695-224EA 408 USD
76695-224 76695-226 77879-983
(2-Ethoxyphenyl)acetic acid 98%
(2-Ethoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: H₃CCH₂O(C₆H₄)CH₂COOH
MDL Number: MFCD00016824
CAS Number: 70289-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR