Diethyl isopropylidenemalonate 95%

Supplier: Ambeed
Warning

A448780-100G A448780-25G A448780-500G A448780-5G
76695-216EA 88 USD
76695-216 76695-218 76695-220 76695-222
Diethyl isopropylidenemalonate 95%
Diethyl isopropylidenemalonate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MDL Number: MFCD00009147
CAS Number: 6802-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR