(R)-(-)-2-Chloromandelic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
A448223-100G
A448223-10G
A448223-25G
A448223-500G
A448223-5G
76695-206EA
59.42
USD
76695-206
76695-208
76695-210
76695-212
76695-214
(R)-(-)-2-Chloromandelic acid 98%
(R)-(-)-2-Chloromandelic acid
Precaución: for Research Use Only
Formula:
C₈H₇ClO₃ MW: 186.59 g/mol |
MDL Number:
MFCD00798429 CAS Number: 52950-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...