Benzyl dimethylphosphonoacetate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethylphosphonoacetic Acid Benzyl Ester, Dimethyl (Cbz-methyl)phosphonate, Benzyl 2-(dimethoxyphosphoryl)acetate, Dimethyl (Benzyloxycarbonylmethyl)phosphonate

A447641-100G A447641-1G A447641-25G A447641-5G
76695-194EA 413.76 USD
76695-194 76695-196 76695-198 76695-200
Benzyl dimethylphosphonoacetate 98%
Benzyl dimethylphosphonoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₅O₅P
MW: 258.21 g/mol
MDL Number: MFCD01872507
CAS Number: 57443-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR