1,2:5,6-Di-O-isopropylidene-α-D-gulofuranose 98%

Supplier: Ambeed
Warning

A444114-100MG A444114-1G A444114-250MG A444114-25G A444114-5G
76695-166EA 7.43 USD
76695-166 76695-168 76695-170 76695-172 76695-174
1,2:5,6-Di-O-isopropylidene-α-D-gulofuranose 98%
1,2:5,6-Di-O-isopropylidene-α-D-gulofuranose

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₆
MW: 260.28 g/mol
MDL Number: MFCD00069693
CAS Number: 14686-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR