Bis(4-methylbenzyl) oxalate 97%

Supplier: Ambeed
Warning

A443059-100G A443059-25G A443059-5G
76695-160EA 50 USD
76695-160 76695-162 76695-164
Bis(4-methylbenzyl) oxalate 97%
Bis(4-methylbenzyl) oxalate

Precaución: for Research Use Only

Formula: C₁₈H₁₈O₄
MDL Number: MFCD00266809
CAS Number: 18241-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR