2-Butylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: sec-Butylboronic acid

A440905-25G A440905-5G
76695-152EA 159.14 USD
76695-152 77551-326
2-Butylboronic acid 96%
2-Butylboronic acid

Precaución: for Research Use Only

Formula: C₄H₁₁BO₂
MW: 101.9400 g/mol
MDL Number: MFCD06656265
CAS Number: 88496-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR