Coumarin-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A440648-100G A440648-100MG A440648-10G A440648-25G A440648-500G A440648-5G
76695-138EA 102.91 USD
76695-138 76695-140 76695-142 76695-144 76695-146 76695-148
Coumarin-3-carboxylic acid 98%
Coumarin-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆O₄
MDL Number: MFCD00006852
CAS Number: 531-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR