2-Phenyl-2-sulfoacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: Alpha-sulphophenylacetic acid

A438489-10G A438489-5G
76695-134EA 18.04 USD
76695-134 77601-908
2-Phenyl-2-sulfoacetic acid 95%
2-Phenyl-2-sulphoacetic acid

Precaución: for Research Use Only

Formula: C₈H₈O₅S
MW: 216.21 g/mol
MDL Number: MFCD09952336
CAS Number: 41360-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR