N-(Carbobenzoxy)aminoacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl (cyanomethyl)carbamate, Cyanomethylcarbamic Acid Benzyl Ester, Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile

A434411-1G A434411-25G A434411-5G
76695-102EA 27 USD
76695-102 76695-104 76695-106
N-(Carbobenzoxy)aminoacetonitrile 98%
N-(Carbobenzoxy)aminoacetonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD00033175
CAS Number: 3589-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR