H-Phe(2-CN)-OH 98%

Supplier: Ambeed
Warning

Synonyms: H-phe(2-cn)-oh

A433209-5G A433209-1G A433209-250MG
76695-084EA 191 USD
76695-084 77878-188 77878-189
H-Phe(2-CN)-OH 98%
H-Phe(2-CN)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD01860884
CAS Number: 263396-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR