Allyl benzyl ether 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Allyl Benzyl Ether, ((Allyloxy)methyl)benzene
A432628-100G
A432628-1G
A432628-25G
A432628-5G
76695-074EA
223
USD
76695-074
76695-076
76695-078
76695-080
Allyl benzyl ether 99%
Allyl benzyl ether
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O |
MDL Number:
MFCD00078288 CAS Number: 14593-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...