6-Hydroxypyridazine-3-carboxylic acid monohydrate 97%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxy-3-pyridazinecarboxylic Acid Monohydrate, 1,6-Dihydro-6-oxopyridazine-3-carboxylic acid monohydrate

A431828-1G A431828-25G A431828-5G
76695-068EA 10.61 USD
76695-068 76695-070 76695-072
6-Hydroxypyridazine-3-carboxylic acid monohydrate 97%
6-Hydroxypyridazine-3-carboxylic acid monohydrate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₄
MW: 158.11 g/mol
MDL Number: MFCD09064936
CAS Number: 306934-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR