Calcium propionate 99%

Supplier: Ambeed
Warning

Synonyms: Calcium dipropionate, Propionic acid hemicalcium salt, Propionic acid calcium salt

A431144-1000G A431144-100G A431144-500G
76695-062EA 16 USD
76695-062 76695-064 76695-066
Calcium propionate 99%
Calcium propionate

Precaución: for Research Use Only

Formula: (CH₃CH₂COO)₂Ca
MW: 186.22 g/mol
MDL Number: MFCD00036137
CAS Number: 4075-81-4
Merck Index: 13,01703

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR