3-Acetoxy-2-methylbenzoic acid 98%

Supplier: Ambeed
Warning

A428387-100G A428387-1G A428387-25G A428387-5G
76695-044EA 272 USD
76695-044 76695-046 76695-048 76695-050
3-Acetoxy-2-methylbenzoic acid 98%
3-Acetoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00957176
CAS Number: 168899-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR