Copper(II) tetrafluoroborate hexahydrate 96%

Supplier: Ambeed
Danger

A428318-5G A428318-25G A428318-10G
76695-042EA 13 USD
76695-042 76842-232 77601-840
Copper(II) tetrafluoroborate hexahydrate 96%
Copper(II) tetrafluoroborate hexahydrate

Precaución: for Research Use Only

Formula: B₂CuF₈H₁₂O₆
MW: 345.243 g/mol
MDL Number: MFCD00150223
CAS Number: 72259-10-0
UN: 3260
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR