Copper(I) trifluoromethanesulfonate benzene complex 98%
Supplier: Ambeed
|
Danger
|
A426791-1G
A426791-5G
A426791-25G
76695-028EA
28
USD
76695-028
76695-030
77879-928
Copper(I) trifluoromethanesulfonate benzene complex 98%
Copper(I) trifluoromethanesulfonate benzene complex
Precaución: for Research Use Only
Formula:
[CuOSO₂CF₃]₂.C₆H₆ |
MDL Number:
MFCD00043300 CAS Number: 42152-46-5 UN: 3175 ADR: 4.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...