(1-Butyl)triphenylphosphonium chloride 98%

Supplier: Ambeed
Warning

Synonyms: Butyltriphenylphosphonium chloride

A426780-1G A426780-100G A426780-25G A426780-5G
76695-026EA 8.49 USD
76695-026 77568-854 77626-850 77626-852
(1-Butyl)triphenylphosphonium chloride 98%
(1-Butyl)triphenylphosphonium chloride

Precaución: For research use only.

Formula: H₃C(CH₂)₃P(C₆H₅)₃Cl
MW: 354.86 g/mol
MDL Number: MFCD00040546
CAS Number: 13371-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR