6-Amino-2-methylquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-methylquinolin-6-amine

A426113-250MG A426113-25G A426113-5G A426113-1G
76695-010EA 30.77 USD
76695-010 76695-012 76695-014 77626-848
6-Amino-2-methylquinoline 98%
6-Amino-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
MDL Number: MFCD00052600
CAS Number: 65079-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR