1,3-Diphenylisobenzofuran 95%

Supplier: Ambeed
Warning

A425113-1G A425113-25G A425113-5G A425113-10G
76694-992EA 18 USD
76694-992 76694-994 76694-996 77878-178
1,3-Diphenylisobenzofuran 95%
1,3-Diphenylisobenzofuran

Precaución: for Research Use Only

Formula: C₂₀H₁₄O
MDL Number: MFCD00005931
CAS Number: 5471-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR